Learn More
Human SLITRK6 Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  R&D Systems AF7317-SP
Product Code. 18744798
  • ONLINE EXCLUSIVE PRICE
    € 209.00
    € 197.40 / 25µg
    Save € 11.60
    5% Off
Description
SLITRK6 Polyclonal specifically detects SLITRK6 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLITRK6
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6
Sheep
Affinity Purified
RUO
84189
Sterile PBS to a final concentration of 0.2 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 2 ug/mL
Q9H5Y7
SLITRK6
Mouse myeloma cell line NS0-derived recombinant human SLITRK6 Ile16-Ala604 Accession # Q9H5Y7
25 μg
Primary
Detects human SLITRK6 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) SLITRK2, rhSLITRK4, and rhSLITRK6 is observed.