missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Slug Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC7408U-100UG
This item is not returnable.
View return policy
Description
Slug Monoclonal specifically detects Slug in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Slug | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGH1, SLUGzinc finger protein, SNAI2, snail 2, snail homolog 2 (Drosophila), SNAIL2, WS2D, zinc finger protein SNAI2 | |
| E. coli-derived recombinant human Slug, Met1-His268, Accession # O43623 | |
| 100 μg | |
| Primary | |
| Detects human Slug in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 666633 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O43623 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6591 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction