Learn More
Human SMAD1 knockout HeLa cell lysate, 1 Kit
Product Code. 32155882 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32155882 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32155882 Supplier Abcam Supplier No. ab2576861Kit
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human SMAD1 knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: SMAD1 knockout HeLa
Specifications
For Use With (Application) Western Blot