missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Smad3 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC4038T-100UG
This item is not returnable.
View return policy
Description
Smad3 Monoclonal specifically detects Smad3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Smad3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P84022 | |
| SMAD3 | |
| E. coli-derived recombinant human Smad3 Ser2-Ala230 Accession # P84022 | |
| RUO | |
| 4088 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 378611 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human Smad3 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction