missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Smad4 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC2097V-100UG
This item is not returnable.
View return policy
Description
Smad4 Monoclonal specifically detects Smad4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Smad4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q13485 | |
| SMAD4 | |
| E. coli-derived recombinant human Smad4 Pro139-Asp332 Accession # Q13485 | |
| RUO | |
| 4089 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 253343 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DPC4, DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Smad4 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction