missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SMPD3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7184-SP
This item is not returnable.
View return policy
Description
SMPD3/NSMase2 Polyclonal specifically detects SMPD3/NSMase2 in Human samples. It is validated for Western Blot.
Specifications
| SMPD3/NSMase2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.4.12, FLJ22593, MGC138443, Neutral sphingomyelinase 2, Neutral sphingomyelinase II, nSMase2, nSMase-2, sphingomyelin phosphodiesterase 3, sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinaseII) | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55512 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q9NY59 | |
| SMPD3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human SMPD3 Val2-Ala655 Accession # Q9NY59 | |
| 25 μg | |
| Primary | |
| Detects human SMPD3 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction