missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX17 NorthernLights™ NL557-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems NL1924R
This item is not returnable.
View return policy
Description
SOX17 Polyclonal specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry.
Specifications
| SOX17 | |
| Polyclonal | |
| Immunocytochemistry | |
| FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3 | |
| Goat | |
| 0.5 mL | |
| Primary | |
| Detects human SOX17 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) SOX2, rhSOX6, and rhSOX15 is observed. | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
| Immunocytochemistry | |
| NorthernLights 557 (NL557) | |
| Q9H6I2 | |
| SOX17 | |
| Affinity Purified | |
| RUO | |
| 64321 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction