missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX2 NorthernLights™ NL493-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems NL20181G
This item is not returnable.
View return policy
Description
SOX2 Polyclonal specifically detects SOX2 in Human samples. It is validated for Immunocytochemistry.
Specifications
| SOX2 | |
| Polyclonal | |
| Immunocytochemistry | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| Goat | |
| 0.5 mL | |
| Primary | |
| Detects human SOX2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SOX17 is observed. | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
| Immunocytochemistry | |
| NorthernLights 493 (NL493) | |
| P48431 | |
| SOX2 | |
| Affinity Purified | |
| RUO | |
| 6657 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction