Learn More
Human SOX9 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 48 publications
Brand: R&D Systems AF3075
This item is not returnable.
View return policy
Description
SOX9 Polyclonal specifically detects SOX9 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| SOX9 | |
| Polyclonal | |
| Unconjugated | |
| P48436 | |
| SOX9 | |
| E. coli-derived recombinant human SOX9 Met1-Lys151 Accession # P48436 | |
| 100 μg | |
| Primary | |
| Detects human SOX9 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human SOX10 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6662 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |