Learn More
Human Sp3 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4256-SP
This item is not returnable.
View return policy
Description
Sp3 Polyclonal specifically detects Sp3 in Human samples. It is validated for Western Blot.
Specifications
| Sp3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686O1631, GC-binding transcription factor Sp3, Sp3 transcription factor, specificity protein 3, SPR2, SPR-2, transcription factor Sp3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6670 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q02447 | |
| SP3 | |
| E. coli-derived recombinant human Sp3 Asp53-Ser309 Accession # Q02447 | |
| 25 μg | |
| Primary | |
| Detects human Sp3 in Western blots. | |
| Human | |
| IgG |