missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPANXC (aa 33-58) Control Fragment Recombinant Protein

Product Code. 30181653
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181653

Brand: Invitrogen™ RP99048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (42%), Rat (42%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83819 (PA5-83819. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NY87
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64663
Name Human SPANXC (aa 33-58) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C; cancer/testis antigen 11.3; cancer/testis antigen family 11, member 3; cancer/testis-associated protein CTp11; cancer/testis-associated protein of 11 kD; cancer-testis-associated protein CTp11; CT11.3; CTp11; nuclear-associated protein SPAN-Xc; SPANX family member C; SPANX family, member C; SPANXC; SPANX-C; SPAN-Xc protein; Sperm protein associated with the nucleus on the x chromosome C; sperm protein associated with the nucleus, x chromosome, family member C
Common Name SPANXC
Gene Symbol SPANXC
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SDPQPAPKKLKTSESSTILVVRYRRN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.