missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SPARC Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC941G-100UG
This item is not returnable.
View return policy
Description
SPARC Monoclonal specifically detects SPARC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| SPARC | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P09486 | |
| SPARC | |
| Mouse myeloma cell line NS0-derived recombinant human SPARC Ala18-Ile303 Accession # P09486 | |
| RUO | |
| 6678 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 122511 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human SPARC/Osteonectin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse SPARC/Osteonectin is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction