missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPEN (aa 139-284) Control Fragment Recombinant Protein

Product Code. 30209354
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209354

Brand: Invitrogen™ RP91596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53388 (PA5-53388. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

May serve as a nuclear matrix platform that organizes and integrates transcriptional responses. In osteoblasts, supports transcription activation: synergizes with RUNX2 to enhance FGFR2-mediated activation of the osteocalcin FGF-responsive element (OCFRE). Has also been shown to be an essential corepressor protein, which probably regulates different key pathways such as the Notch pathway. Negative regulator of the Notch pathway via its interaction with RBPSUH, which prevents the association between NOTCH1 and RBPSUH, and therefore suppresses the transactivation activity of Notch signaling. Blocks the differentiation of precursor B-cells into marginal zone B-cells. Probably represses transcription via the recruitment of large complexes containing histone deacetylase proteins. May bind both to DNA and RNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96T58
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23013
Name Human SPEN (aa 139-284) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HIAA0929; KIAA0929; MINT; Msx2 interacting nuclear target (MINT) homolog; msx2-interacting protein; nuclear receptor transcription cofactor; RBM15C; SHARP; SMART/HDAC1-associated repressor protein; SPEN; spen family transcriptional repressor; SPEN homolog; spen homolog, transcriptional regulator
Common Name SPEN
Gene Symbol SPEN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NRERAYEHSAYGHHERGTGGFDRTRHYDQDYYRDPRERTLQHGLYYASRSRSPNRFDAHDPRYEPRAREQFTLPSVVHRDIYRDDITREVRGRRPERNYQHSRSRSPHSSQSRNQSPQRLASQASRPTRSPSGSGSRSRSSSSDSI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.