missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SPINK1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7496
This item is not returnable.
View return policy
Description
SPINK1 Polyclonal specifically detects SPINK1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| SPINK1 | |
| Polyclonal | |
| Unconjugated | |
| P00995 | |
| SPINK1 | |
| E. coli-derived recombinant human SPINK1 Asp24-Cys79 Accession # P00995 | |
| 100 μg | |
| Primary | |
| Detects human SPINK1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
| pancreatic secretory trypsin inhibitor, PCTT, PCTTSpink3, PSTI, PSTISerine protease inhibitor Kazal-type 1, serine peptidase inhibitor, Kazal type 1, serine protease inhibitor, Kazal type 1, Spink3, TATI, TATITumor-associated trypsin inhibitor | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6690 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction