missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ST3GAL6 Synthetic Peptide

Product Code. 15339100
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 15339100

Brand: Invitrogen™ PEP0862

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

ST3gal6 Synthetic Peptide, PEP-0862, from Invitrogen.

Sialyltransferases catalyze the transfer of sialic acid from cytidine 5-prime monophospho-N-acetylneuraminic acid (CMP-NeuAc) to terminal positions of glycoprotein and glycolipid carbohydrate groups. Terminal NeuAc residues are key determinants of carbohydrate structures, such as the sialyl-Lewis X determinants, and are widely distributed in many cell types. However, cancer cells often express more heavily sialylated glycans on their cell surface and this feature sometimes correlates with invasiveness. In contrast, expression of ST3gal6, a member of the sialyltransferase family that sialylates type II lactosamine structures on glycoproteins and glycolipids, was found to be significantly decreased by hypermethylation of the gene in gastrointestinal cancer. At least three isoforms of ST3gal6 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 10402
Species Human
Purification Method purified
Accession Number Q9Y274
Common Name Human ST3GAL6
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ST3GAL6
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.