missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ST7/LRP12 Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems BAF2560
This item is not returnable.
View return policy
Description
ST7/LRP12 Polyclonal specifically detects ST7/LRP12 in Human samples. It is validated for Western Blot.
Specifications
| ST7/LRP12 | |
| Polyclonal | |
| Biotin | |
| Q9Y561 | |
| LRP12 | |
| Mouse myeloma cell line NS0-derived recombinant human ST7/LRP12 Asn28-Ile488 Accession # Q9Y561 | |
| 50 μg | |
| Primary | |
| Detects human ST7/LRP12 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| DKFZp762O2113, FAM4A1, LRP12, Protein FAM4A1, Protein HELG, RAY1, SEN4, subfamily A, member 1, suppression of tumorigenicity 7, suppression of tumorigenicity 7 (breast), suppressor of tumorigenicity 7 protein, TSG7 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 29967 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction