missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STAMP2/STEAP4 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4626U-100UG
This item is not returnable.
View return policy
Description
STAMP2/STEAP4 Monoclonal specifically detects STAMP2/STEAP4 in Human samples. It is validated for Flow Cytometry.
Specifications
| STAMP2/STEAP4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| EAW76907 | |
| STEAP4 | |
| NS0 mouse myeloma cell line transfected with human STAMP2 Met1-His459 Accession # EAW76907 | |
| RUO | |
| 79689 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 418714 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 1.16.1, EC 1.16.1.-, FLJ23153, Six-transmembrane epithelial antigen of prostate 4, SixTransMembrane protein of prostate 2, STAMP2DKFZp666D049, STEAP family member 4, STEAP4, TIARP, TIARPsix transmembrane prostate protein 2, TNFAIP9, Tumor necrosis factor, alpha-induced protein 9metalloreductase STEAP4, tumor necrosis-alpha-induced adipose-related protein | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human STAMP2-transfected NS0 cells but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction