missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STAT5a Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC21741U-100UG
This item is not returnable.
View return policy
Description
STAT5A Monoclonal specifically detects STAT5A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| STAT5a | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_003143 | |
| STAT5A | |
| Human STAT5a synthetic peptide SLDSRLSPPAGLFTSARGSLS Accession # NP_003143 | |
| RUO | |
| 6776 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 251610 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MGF, signal transducer and activator of transcription 5A, STAT5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human STAT5a. This antibody does not cross-react with STAT5b. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction