missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STAT5a/b Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC41901S-100UG
This item is not returnable.
View return policy
Description
STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| STAT5a/b | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694), Accession # P51692 | |
| 100 μg | |
| Primary | |
| Detects human STAT5a/b when phosphorylated at Y964/Y699 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
| Flow Cytometry | |
| 1247C | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P51692 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6776 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction