Learn More
Invitrogen™ TNFR1 (Soluble-60 kDa) Human ELISA Kit
For the quantitative detection of human sTNF-R (60 kd).
€ 634.00 - € 4325.00
Specifications
Accession Number | P19438 |
---|---|
Assay Range | 0.08-5.0 ng/mL |
Assay Sensitivity | 0.05 ng/mL |
Conjugate | HRP |
Product Type | ELISA Kit |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal polyclonal antibody to human sTNF-R (60 kDa)
HRP-Conjugate anti-human sTNF-R (60 kDa) monoclonal antibody
Human sTNF-R (60 kDa) Standard, 10ng/mL upon dilution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15541527
|
Invitrogen™
BMS203 |
96 Tests |
€ 634.00
96 tests |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
15551547
|
Invitrogen™
BMS203TEN |
10 x 96 Tests |
€ 4325.00
Pack of 10 |
Estimated Shipment: 01-05-2024
Log in to see stock. |
|||||
Description
The Human Soluble Tumor Necrosis Factor Receptor 1 (sTNFR1-60 kDa) ELISA quantitates sTNFR1 in human serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu sTNFR1. Principle of the method The Human sTNFR1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
- Provides a simple, rapid and highly sensitive method for the determination of soluble TNF-R (60 kd) levels in body fluids or cell culture supernatants
- This assay will help to clarify the possible diagnostic and prognostic value of circulating sTNF-R (60 kd) in various neoplastic and inflammatory diseases
Specifications
P19438 | |
0.05 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
CD120a,FPF,TBP1,TNF-R,TNF-R-I,TNF-R55,TNFAR,TNFR1,TNFR55,TNFR60,p55,p55-R,p60 | |
1.9% | |
HRP | |
Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL; Urine,10 μL | |
Tumor necrosis factor receptor 1, TNF-R1, CD120a | |
1 hr. 20 min. |
0.08-5.0 ng/mL | |
HRP | |
Plasma, Serum, Supernatant, Urine | |
ELISA | |
7132 | |
8.6% | |
Pre-coated 96 well plates, Standard, Sample Diluent, Assay Buffer concentrate, Detection Antibody Conjugate: anti-rabbit-HRP, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
Human | |
2 hr. 10 min. |
For Research Use Only