missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human STS1 (aa 297-370) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144780 (PA5-144780. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Sts1 encodes a protein that contains a ubiquitin associated domain at the N-terminus, an SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. The encoded protein was found to inhibit endocytosis of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor.
Specifications
Specifications
| Accession Number | Q8TF42 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 84959 |
| Name | Human STS1 (aa 297-370) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2810457I06Rik; BB125008; Cbl-interacting protein p70; Cbl-interacting protein Sts-1; KIAA1959; MGC15437; nm23-phosphorylated unknown substrate; p70; RGD1310357; SH3 domain-containing 70 kDa protein, suppressor of T-cell receptor signaling 1, nm23-phosphorylated unknown substrate; STS1; STS-1; suppressor of T-cell receptor signaling 1; T-cell ubiquitin ligand 2; TULA2; TULA-2; tyrosine-protein phosphatase STS1/TULA2; UBASH3B; ubiquitin associated and SH3 domain containing B; ubiquitin associated and SH3 domain containing, B; ubiquitin-associated and SH3 domain-containing protein B |
| Common Name | STS1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction