missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Survivin Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC886T-100UG
This item is not returnable.
View return policy
Description
Survivin Monoclonal specifically detects Survivin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Survivin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| E. coli-derived recombinant human Survivin Met1-Asp142 Accession # O15392 | |
| RUO | |
| 332 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 91630 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O15392 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Survivin in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction