missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SUSD2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB90562U-100UG
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Flow Cytometry.
Specifications
| SUSD2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| HEK293 human embryonic kidney cell line-derived human SUSD2 Met1-Ala785 Accession # Q9UGT4 | |
| RUO | |
| 56241 | |
| Human | |
| IgG |
| Flow Cytometry | |
| 1279A | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9UGT4 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects human SUSD2 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction