Learn More
Human T-bet/TBX21 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB53851U-100UG
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9UL17 | |
| TBX21 | |
| E. coli-derived recombinant human T-bet/TBX21 Glu326-Asn535 Accession # Q9UL17 | |
| 100 μg | |
| Primary | |
| Detects human T-bet/TBX21 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 525831 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Tbet, T-bet, TBETT-PET, TBLYM, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, TBX21, T-PET, Transcription factor TBLYM | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 30009 | |
| Human | |
| Purified |
Product Content Correction
Product Title
For Research Use Only (RUO)