missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACE/ADAM17 Ectodomain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB930-SP
This item is not returnable.
View return policy
Description
TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot.
Specifications
| TACE/ADAM17 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6868 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 111636 | |
| Western Blot 1 ug/mL | |
| P78536 | |
| ADAM17 | |
| Insect ovarian cell line T. ni-derived recombinant human TACE/ADAM17 Pro18-Asn671 (predicted) Accession # P78536 | |
| 25 μg | |
| Primary | |
| Detects TACE/ADAM17 Ectodomain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with the ectodomain of recombinant human (rh) ADAM8, rhADAM9, or rhADAM15 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction