missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TAFA2/FAM19A2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4179-SP
This item is not returnable.
View return policy
Description
TAFA2/FAM19A2 Polyclonal specifically detects TAFA2/FAM19A2 in Human samples. It is validated for Immunohistochemistry, Neutralization.
Specifications
| TAFA2/FAM19A2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Chemokine-like protein TAFA-2, DKFZp761E1217, DKFZp781P0552, FAM19A2, family with sequence similarity 19 (chemokine (C-C motif)-like), member A2, MGC42403, TAFA2, TAFA-2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 338811 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry, Neutralization | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL, Neutralization 5 ug/mL | |
| Q8N3H0 | |
| FAM19A2 | |
| Mouse myeloma cell line NS0-derived recombinant human TAFA2/FAM19A2 Ala31-His131 Accession # Q8N3H0 | |
| 25 μg | |
| Primary | |
| Detects human TAFA2/FAM19A2 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human TAFA5 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction