missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TAFA5/FAM19A5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5148-SP
This item is not returnable.
View return policy
Description
TAFA5/FAM19A5 Monoclonal specifically detects TAFA5/FAM19A5 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TAFA5/FAM19A5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| NP_056196 | |
| FAM19A5 | |
| E. coli-derived recombinant human TAFA5/FAM19A5 Gln26-Ser125 Accession # NP_056196 | |
| 25 μg | |
| Primary | |
| Detects TAFA5/FAM19A5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA2, 3, or 4 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 463102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Chemokine-like protein TAFA-5, FAM19A5, family with sequence similarity 19 (chemokine (C-C motif)-like), member A5, QLLK5208, TAFA protein 5, TAFA5, TAFA-5, UNQ5208 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25817 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction