missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TAP2 Antibody, R&D Systems™

Description
TAP2 Monoclonal specifically detects TAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
Specifications
| Antigen | TAP2 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 998524 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 μg/mL, Immunohistochemistry 8-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q03519 |
| Gene Alias | ABC18, ABCB3, ABCB3ABC18, antigen peptide transporter 2, APT2, ATP-binding cassette, sub-family B (MDR/TAP), member 3, D6S217E, MHC 2, Peptide transporter involved in antigen processing 2, Peptide transporter PSF2, Peptide transporter TAP2, PSF2, PSF2PSF-2, Really interesting new gene 11 protein, RING11, RING11ATP-binding cassette sub-family B member 3, transporter 2, ABC (ATP binding cassette), transporter 2, ATP-binding cassette, sub-family B (MDR/TAP), Y1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?