Learn More
Human TAPBPR Alexa Fluor™ 700-conjugated Antibody, Novus Biologicals™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB11337N-100UG
Product Code. 18705825
  • € 618.00 / 100µg
Description
Tapasin Related Protein Monoclonal antibody specifically detects Tapasin Related Protein in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tapasin Related Protein
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBPL, TAPBPR, TAPBP-RTAP binding protein related, TAPBPRtapasin-like
Chinese Hamster Ovary cell line, CHO-derived human TAPBPR, Ala19-Arg404, Accession # Q9BX59.2
100 μg
Primary
Human
Purified
Flow Cytometry
1059329
Flow Cytometry
Q9BX59.2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
55080
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
IgG2a