missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TBX4 (aa 265-382) Control Fragment Recombinant Protein

Product Code. 30204439
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204439

Brand: Invitrogen™ RP106069

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65383 (PA5-65383. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is the human homolog of mouse Tbx4, which is closely linked to Tbx2 on mouse chromosome 11. Similarly this gene, like TBX2, maps to human chromosome 17. Expression studies in mouse and chicken show that Tbx4 is expressed in developing hindlimb, but not in forelimb buds, suggesting a role for this gene in regulating limb development and specification of limb identity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P57082
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9496
Name Human TBX4 (aa 265-382) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3930401C23; ICPPS; SPS; T-box 4; T-box 4 protein; T-box protein 4; T-box transcription factor 4; T-box transcription factor TB x 4; Tb x 4; transcription factor Tb x 4
Common Name TBX4
Gene Symbol TBX4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KEYPVISKSIMRQRLISPQLSATPDVGPLLGTHQALQHYQHENGAHSQLAEPQDLPLSTFPTQRDSSLFYHCLKRRADGTRHLDLPCKRSYLEAPSSVGEDHYFRSPPPYDQQMLSPS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.