missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TCF-3/E2A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB44062-SP
This item is not returnable.
View return policy
Description
TCF-3/E2A Monoclonal specifically detects TCF-3/E2A in Human samples. It is validated for Western Blot.
Specifications
| TCF-3/E2A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| NP_003191 | |
| TCF3 | |
| E.coli-derived recombinant human TCF-3/E2A Gln100-Glu500 Accession # NP_003191 | |
| 25 μg | |
| Primary | |
| Detects human TCF-3/E2A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TCF-1 (aa 116-233), rhTCF-1 (aa 217-358), rhTCF-1 alpha/LEF-1, rhTCF-2, rhTCF-4, rhTCF-12, rhTCF7L1 (aa 151-320), rhTCFL1 (aa 429-581), or rhTCF7L2 (aa 131-300) is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 401703 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BHLHB21, bHLHb21TCF-3, Class B basic helix-loop-helix protein 21, E2A, E2A immunoglobulin enhancer-binding factor E12/E47, E2AKappa-E2-binding factor, Immunoglobulin enhancer-binding factor E12/E47, Immunoglobulin transcription factor 1, ITF1, ITF1Transcription factor ITF-1, Kappa-E2-binding factor, MGC129647, MGC129648, Pan2, Tcfe2a, Transcription factor 3, transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47), transcription factor E2-alpha, VDIR, VDR interacting repressor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6929 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction