missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Tenascin R ELISA Kit

Product Code. 17707803
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17707803

Brand: Invitrogen™ EH447RBX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Tenascin-R (TNR) is an extracellular matix protein expressed primarily in the central nervous system. It is a member of the tenascin (TN) gene family, which includes at least 3 genes in mammals: TNC (or hexabrachion; MIM 187380), TNX (TNXB; MIM 600985), and TNR (Erickson, 1993). The genes are expressed in distinct tissues at different times during embryonic development and are present in adult tissues.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92752
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7143
Gene Symbol Tnr
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Tenascin R
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.