missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TfR (Transferrin R) Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF2474-SP
This item is not returnable.
View return policy
Description
TfR (Transferrin R) Polyclonal specifically detects TfR (Transferrin R) in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifications
| TfR (Transferrin R) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TFRC | |
| Human placenta-derived TfR (Transferrin R) | |
| 25 μg | |
| Primary | |
| Detects human TfR (Transferrin R) in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
| CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7037 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction