missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TGR5/GPBAR1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4286T-100UG
This item is not returnable.
View return policy
Description
TGR5/GPBAR1 Monoclonal specifically detects TGR5/GPBAR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| TGR5/GPBAR1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8TDU6 | |
| GPBAR1 | |
| NS0 mouse myeloma cell line transfected with human TGR5/GPBAR1 Met1-Asn330 Accession # Q8TDU6 | |
| 100 μg | |
| Primary | |
| Stains human TGR5/GPBAR1 transfectants but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
| Flow Cytometry | |
| 409522 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BG37, BG37hBG37, G protein-coupled bile acid receptor 1, GPBA, GPBAR1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BAR, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 151306 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction