missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human Th17 Cytokine Staining Panel

Product Code. 15571227
Click to view available options
Quantity:
1 panel
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15571227

Brand: Invitrogen™ 88841944

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Anti-Human IL-17A (eBio64Dec17) FITC : 25 tests (0.125 μg/5 μL test)*
  • Anti-Human IL-17F (SHLR17) PE : 25 tests (0.5 μg/5 μL test)*
  • Anti-Human IL-22 (22URTI) PerCP-eFluor 710 : 25 tests (0.015 μg/5 μL test)*
  • Anti-Human IL-21 (3A3-N2) eFluor 660 : 25 tests (0.125 μg/5 μL test)*
  • Anti-Human CD4 (RPA-T4) eFluor 450 : 25 tests (0.25 μg/5 μL test)*
  • IC Fixation Buffer
  • Permeabilization Buffer (10x)
  • *A test is defined as the amount of antibody to use for 1 million cells in a 100 μL staining volume.

Includes all reagents needed for simultaneous flow cytometric detection of all the major cytokines produced by the Th17 lineage (IL-17A, IL-17F, IL-21 and IL-22).

  • 5μL/test
  • Also included: An anti-CD4 antibody that can be used after fixation and permeabilization, as well as IC Fixation and Permeabilization Buffers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C. Light sensitive material.
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 1 panel
For Use With (Application) Intracellular Staining, Flow Cytometry
Product Line eBioscience
Product Type Staining Panel

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.