missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Thrombospondin-1 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
1 set
Description
The Human Thrombospondin-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Thrombospondin-1.
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 1.56 to 100 ng/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7057 |
| Synonym | THBS, THBS1, THBS-1, thrombospondin 1, Thrombospondin1, thrombospondin-1p180, TSP-1, TSP1thrombospondin-1, TSPTSP-1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction