missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Thrombospondin-2 Quantikine ELISA Kit

Product Code. 18782054
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18782054

Brand: R&D Systems™ DTSP20

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Thrombospondin-2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Thrombospondin-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Thrombospondin-2 in cell culture supernates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul), human milk (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.31 - 20ng/mL
Assay Sensitivity 0.068ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human Thrombospondin-2
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 7058
Sample Volume Cell Culture Supernates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym THBS2, thrombospondin 2, Thrombospondin2, TSP-2, TSP2thrombospondin-2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.