missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tie-1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB619P
This item is not returnable.
View return policy
Description
Tie-1 Polyclonal specifically detects Tie-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Tie-1 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, Tie1, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7075 | |
| Human | |
| IgG |
| Flow Cytometry | |
| R-PE | |
| P35590 | |
| TIE1 | |
| Mouse myeloma cell line NS0-derived recombinant human Tie-1 Ala22-Gln760 Accession # P35590 | |
| 100 Tests | |
| Primary | |
| Detects human Tie-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Tie-2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction