missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-1/KIM-1/HAVCR Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB17504G-100UG
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Monoclonal antibody specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for Flow Cytometry
Specifications
| TIM-1/KIM-1/HAVCR | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| Mouse myeloma cell line NS0-derived human TIM-1/KIM-1/HAVCR, Pro21-Thr288, Accession # AAC39862 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2389F | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| AAC39862 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26762 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction