missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-1/KIM-1/HAVCR Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1750N-100UG
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for Flow Cytometry.
Specifications
| TIM-1/KIM-1/HAVCR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAC39862 | |
| HAVCR1 | |
| Mouse myeloma cell line NS0-derived recombinant human TIM-1/KIM-1/HAVCR Ser21-Thr288 Accession # AAC39862 | |
| RUO | |
| 26762 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 219211 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TIM-1/KIM-1/HAVCR in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) TIM-1, rmTIM-2, or rhTIM-3. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction