missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-3 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB23652U-100UG
This item is not returnable.
View return policy
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry.
Specifications
| TIM-3 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
| Mouse myeloma cell line NS0-derived recombinant human TIM-3, Met1-Arg200, Accession No. Q8TDQ0 | |
| 100 μg | |
| 84868.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2321C | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q8TDQ0 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human TIM-3 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction