missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIM-4 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2929RT-100UG
This item is not returnable.
View return policy
Description
TIM-4 Monoclonal specifically detects TIM-4 in Human samples. It is validated for Flow Cytometry.
Specifications
| TIM-4 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
| FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD4, TIMD-4 | |
| Mouse myeloma cell line NSO-derived recombinant human TIM-4 Glu25-Leu315 Accession # Q96H15 | |
| 100 μg | |
| 91937 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 921832R | |
| Flow Cytometry 0.25-1 μg/10^6 cells | |
| Q96H15 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human TIM-4 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction