missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TIMM13 (aa 1-75) Control Fragment Recombinant Protein

Product Code. 30199999
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199999

Brand: Invitrogen™ RP101141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61856 (PA5-61856. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the evolutionarily conserved TIMM (translocase of inner mitochondrial membrane) family of proteins that function as chaperones in the import of proteins from the cytoplasm into the mitochondrial inner membrane. Proteins of this family play a role in collecting substrate proteins from the translocase of the outer mitochondrial membrane (TOM) complex and delivering them to either the sorting and assembly machinery in the outer mitochondrial membrane (SAM) complex or the TIMM22 complex in the inner mitochondrial membrane. The encoded protein and the translocase of mitochondrial inner membrane 8a protein form a 70 kDa complex in the intermembrane space.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y5L4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26517
Name Human TIMM13 (aa 1-75) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias D10Ertd378e; fd44h11; hypothetical protein LOC436738; mitochondrial import inner membrane translocase subunit Tim13; mitochondrial import inner membrane translocase subunit Tim13B; ppv1; TIM13; Tim13a; TIM13B; Tim9; Timm13; TIMM13A; TIMM13B; Timm9; translocase of inner mitochondrial membrane 13; translocase of inner mitochondrial membrane 13 homolog; translocase of inner mitochondrial membrane 13 homolog (yeast); translocase of inner mitochondrial membrane 9 homolog; wu:fd44h11; zgc:92895
Common Name TIMM13
Gene Symbol TIMM13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MEGGFGSDFGGSGSGKLDPGLIMEQVKVQIAVANAQELLQRMTDKCFRKCIGKPGGSLDNSEQKCIAMCMDRYMD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.