missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIMP-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 13 publications
Brand: R&D Systems AF970
This item is not returnable.
View return policy
Description
TIMP-1 Polyclonal specifically detects TIMP-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization, Knockout Validated.
Specifications
| TIMP-1 | |
| Polyclonal | |
| Unconjugated | |
| Q6FGX5 | |
| TIMP1 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-1 Cys24-Ala207 Accession # Q6FGX5 | |
| 100 μg | |
| Primary | |
| Detects human TIMP-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse TIMP-1 and recombinant rat TIMP-1 is observed and less than 1% cross-reactivity with recombinant human (rh) TIMP-2, rhTIMP-3, and rhTIMP-4 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization, Immunoassay | |
| LYOPH | |
| Western Blot 1-5 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 1-15 ug/mL, Neutralization 1 ug/mL, Knockout Validated | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7076 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction