missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TIMP-3 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC9731R-100UG
This item is not returnable.
View return policy
Description
TIMP-3 Monoclonal specifically detects TIMP-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TIMP-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P35625 | |
| TIMP3 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-3 Cys24-Pro211 Accession # P35625 | |
| RUO | |
| 7078 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 277128 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TIMP-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TIMP-1, -2, or -4 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction