missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TLR9 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC36583R-100UG
This item is not returnable.
View return policy
Description
TLR9 Monoclonal specifically detects TLR9 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TLR9 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| Antibody developed against the KLH-conjugated synthetic peptide corresponding to amino acids Asp250-Asn300 of human TLR9 isoform A., Accession No. Q9NR96 | |
| 100 μg | |
| 54106.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 26C593R | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| Q9NR96 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human TLR9 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction