missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMEM87A Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7966N-100UG
This item is not returnable.
View return policy
Description
TMEM87A Monoclonal specifically detects TMEM87A in Human samples. It is validated for Flow Cytometry.
Specifications
| TMEM87A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8NBN3 | |
| TMEM87A | |
| E. coli-derived recombinant human TMEM87A Ser22-Asn157 Accession # Q8NBN3 | |
| RUO | |
| 25963 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 772807 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| DKFZp564G2022, FLJ11273, MGC33727, PSEC0094, transmembrane protein 87A | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TMEM87A in ELISAs and Western Blots | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction