missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TNF-alpha XL Luminex™ Performance Assay

Product Code. 30001512
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
100 tests
Product Code. Quantity unitSize
30001512 1 Kit 100 tests
1 options
This item is not returnable. View return policy

Product Code. 30001512

Brand: R&D Systems™ LUXLM210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target TNF-alpha
Gene ID (Entrez) 7124
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.