missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNFRSF10A (1 a.a. - 238 a.a.) from HEK 293 cells with human IgG1 Recombinant Protein

Product Code. 16272180
Click to view available options
:
50μg
This item is not returnable. View return policy

Product Code. 16272180

Brand: Abnova™ P5031.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10/TRAIL), and thus transduces cell death signal and induces cell apoptosis. Studies with FADD-deficient mice suggested that FADD, a death domain containing adaptor protein, is required for the apoptosis mediated by this protein. [provided by RefSeq]

Specifications

For Use With (Application) SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 8797
Molecular Weight (g/mol) 43kDa
Name TNFRSF10A (Human) Recombinant Protein with Fc
Purification Method Protein A purification
Quality Control Testing 12% SDS-PAGE SDS-PAGE Stained with Coomassie Blue
Quantity 50 ug
Storage Requirements Store at -20°C on dry atmosphere for at least 6 months. After reconstitution with 50 uL sterile deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.2 EU/μg purified protein as determined by LAL test
Gene Alias APO2/CD261/DR4/MGC9365/TRAILR-1/TRAILR1
Common Name TNFRSF10A
Gene Symbol TNFRSF10A
Species Human
Recombinant Recombinant
Protein Tag None
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.