missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNFRSF4 Recombinant Protein fused to murine IgG2a Fc purifird from CHO cells

Product Code. 16141680
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 16141680

Brand: Abnova™ P4934.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB through its interaction with adaptor proteins TRAF2 and TRAF5. Knockout studies in mice suggested that this receptor promotes the expression of apoptosis inhibitors BCL2 and BCL2lL1/BCL2-XL, and thus suppresses apoptosis. The knockout studies also suggested the roles of this receptor in CD4+ T cell response, as well as in T cell-dependent B cell proliferation and differentiation. [provided by RefSeq]

Sequence: TNFRSF4 mature: lhcvgdtypsndrcchecrpgngmvsrcsrsqntvcrpcgpgfyndvvsskpckpctwcnlrsgserkqlctatqdtvcrcragtqpldsykpgvdcapcppghfspgdnqackpwtnctlagkhtlqpasnssdaicedrdppatqpqetqgpparpitvqpteawprtsqgpstr

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 7293
Molecular Weight (g/mol) 45.6kDa
Name TNFRSF4 (Human) Recombinanat Protein
Preparation Method Mammalian cell (CHO) expression system
Purification Method Size purification
Quantity 25 μg
Immunogen TNFRSF4 mature: lhcvgdtypsndrcchecrpgngmvsrcsrsqntvcrpcgpgfyndvvsskpckpctwcnlrsgserkqlctatqdtvcrcragtqpldsykpgvdcapcppghfspgdnqackpwtnctlagkhtlqpasnssdaicedrdppatqpqetqgpparpitvqpteawprtsqgpstr Fused to murine IgG2a Fc: eprgptikpcppckcpapnllggpsvfifppkikdvlmislspivtcvvvdvseddpdvqiswfvnnvevhtaqtqthredynstlrvvsalpiqhqdwmsgkefkckvnnkdlpapiertiskpkgsvrapqvyvlpppeeemtkkqvtltcmvtdfmpediyvewtnngktelnykntepvldsdgsyfmysklrvekknwvernsyscsvvheglhnhhttksfsrtpg
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Gene Alias ACT35/CD134/OX40/TXGP1L
Common Name TNFRSF4
Gene Symbol TNFRSF4
Species Human
Recombinant Recombinant
Protein Tag None
Expression System Mammalian cell (CHO) expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.